2,3-Dichloromaleic anhydride, 97%
2,3-Dichloromaleic anhydride, 97%
2,3-Dichloromaleic anhydride, 97%
Thermo Scientific Chemicals

2,3-Dichloromaleic anhydride, 97%

CAS: 1122-17-4 | C4Cl2O3 | 166.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 113440050
Price (EUR)
54,30
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
54,30
Each
Specifications
Assay Percent Range96% min. (GC)
Beilstein17,434
CAS1122-17-4
Chemical Name or Material2,3-Dichloromaleic anhydride
EINECS Number214-343-
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only