2,3-Dimercapto-1-propanol, 97.0-100.5%
2,3-Dimercapto-1-propanol, 97.0-100.5%
2,3-Dimercapto-1-propanol, 97.0-100.5%
Thermo Scientific Chemicals

2,3-Dimercapto-1-propanol, 97.0-100.5%

CAS: 59-52-9 | C3H8OS2 | 124.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 115300250
Price (EUR)
478,65
Online Exclusive
532,00
Save 53,35 (10%)
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
478,65
Online Exclusive
532,00
Save 53,35 (10%)
Each
Chemical Identifiers
CAS59-52-9
IUPAC Name2,3-disulfanylpropan-1-ol
Molecular FormulaC3H8OS2
InChI KeyWQABCVAJNWAXTE-UHFFFAOYNA-N
SMILESOCC(S)CS
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5700 to 1.5750 (20°C, 589 nm)
Titration Iodimetric97.0 to 100.5 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only