3,5-Dimethoxybenzoic acid, 98%
3,5-Dimethoxybenzoic acid, 98%
3,5-Dimethoxybenzoic acid, 98%
Thermo Scientific Chemicals

3,5-Dimethoxybenzoic acid, 98%

CAS: 1132-21-4 | C9H10O4 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 115461000
Price (EUR)
150,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
150,00
Each
Chemical Identifiers
CAS1132-21-4
IUPAC Name3,5-dimethoxybenzoic acid
Molecular FormulaC9H10O4
InChI KeyIWPZKOJSYQZABD-UHFFFAOYSA-N
SMILESCOC1=CC(=CC(OC)=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point182°C to 185°C
Titration with NaOH97.5 to 102.5 %
HPLC>=97.5 %
Appearance (Color)White to pink-beige
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only