2-Dimethylaminoethyl chloride hydrochloride, 99%, Thermo Scientific Chemicals
2-Dimethylaminoethyl chloride hydrochloride, 99%, Thermo Scientific Chemicals
2-Dimethylaminoethyl chloride hydrochloride, 99%, Thermo Scientific Chemicals
2-Dimethylaminoethyl chloride hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Dimethylaminoethyl chloride hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 115825000
Price (EUR)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS4584-46-7
IUPAC Namehydrogen (2-chloroethyl)dimethylamine chloride
Molecular FormulaC4H11Cl2N
InChI KeyLQLJZSJKRYTKTP-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)CCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Ionic chloride24.1 to 25.1 %
Total chloride48.4 to 50.0 %
Ionic chloride24.1 to 25.1 %
Total chloride48.4 to 50.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only