3,3-Dimethylglutaric acid, 98%
3,3-Dimethylglutaric acid, 98%
3,3-Dimethylglutaric acid, 98%
Thermo Scientific Chemicals

3,3-Dimethylglutaric acid, 98%

CAS: 4839-46-7 | C7H12O4 | 160.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 116241000
Price (EUR)
144,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
144,00
Each
Chemical Identifiers
CAS4839-46-7
IUPAC Name3,3-dimethylpentanedioic acid
Molecular FormulaC7H12O4
InChI KeyDUHQIGLHYXLKAE-UHFFFAOYSA-N
SMILESCC(C)(CC(O)=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Melting point98°C to 102°C
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only