2,3-Dimethylnaphthalene, 97%
2,3-Dimethylnaphthalene, 97%
2,3-Dimethylnaphthalene, 97%
Thermo Scientific Chemicals

2,3-Dimethylnaphthalene, 97%

CAS: 581-40-8 | C12H12 | 156.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 116450050
Price (EUR)
193,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
193,00
Each
Chemical Identifiers
CAS581-40-8
IUPAC Name2,3-dimethylnaphthalene
Molecular FormulaC12H12
InChI KeyWWGUMAYGTYQSGA-UHFFFAOYSA-N
SMILESCC1=C(C)C=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point102°C to 106°C
Appearance (Color)Off-white to cream
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only