2,6-Dimethyl-gamma-pyrone, 99%
2,6-Dimethyl-gamma-pyrone, 99%
2,6-Dimethyl-gamma-pyrone, 99%
Thermo Scientific Chemicals

2,6-Dimethyl-gamma-pyrone, 99%

CAS: 1004-36-0 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 116741000
Price (EUR)
461,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
461,00
Each
Chemical Identifiers
CAS1004-36-0
IUPAC Name2,6-dimethyl-4H-pyran-4-one
Molecular FormulaC7H8O2
InChI KeyVSYFZULSKMFUJJ-UHFFFAOYSA-N
SMILESCC1=CC(=O)C=C(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to beige
Melting point132°C to 137°C
GC>=98.5 %
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only