Ethylbenzene, 99.8%, pure
Ethylbenzene, 99.8%, pure
Ethylbenzene, 99.8%, pure
Thermo Scientific Chemicals

Ethylbenzene, 99.8%, pure

CAS: 100-41-4 | C8H10 | 106.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
500 mL
1 L
2.5 L
25 L
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 118080010
Price (EUR)
34,65
Online exclusive
38,00
Save 3,35 (9%)
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
34,65
Online exclusive
38,00
Save 3,35 (9%)
Each
Chemical Identifiers
CAS100-41-4
IUPAC Nameethylbenzene
Molecular FormulaC8H10
InChI KeyYNQLUTRBYVCPMQ-UHFFFAOYSA-N
SMILESCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Refractive index1.4940 to 1.4970 (20°C, 589 nm)
Infrared spectrumConforms
Water=<0.02 % (Coulometric)
Color scale=<10 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only