Fluorescein Isothiocyanate Isomer I, 90%, Pure
Fluorescein Isothiocyanate Isomer I, 90%, Pure
Fluorescein Isothiocyanate Isomer I, 90%, Pure
Thermo Scientific Chemicals

Fluorescein Isothiocyanate Isomer I, 90%, Pure

CAS: 3326-32-7 | C21H11NO5S | 389.381 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number 119250050
Price (EUR)
994,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
994,00
Each
Chemical Identifiers
CAS3326-32-7
IUPAC Name3',6'-dihydroxy-5-isothiocyanato-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular FormulaC21H11NO5S
InChI KeyMHMNJMPURVTYEJ-UHFFFAOYSA-N
SMILESOC1=CC=C2C(OC3=CC(O)=CC=C3C22OC(=O)C3=CC(=CC=C23)N=C=S)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Infrared spectrumConforms
HPLC>=88 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only