2-Fluorobenzoic acid, 99%
2-Fluorobenzoic acid, 99%
2-Fluorobenzoic acid, 99%
Thermo Scientific Chemicals

2-Fluorobenzoic acid, 99%

CAS: 445-29-4 | C7H5FO2 | 140.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 119360250
Price (EUR)
25,40
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
25,40
Each
Chemical Identifiers
CAS445-29-4
IUPAC Name2-fluorobenzoic acid
Molecular FormulaC7H5FO2
InChI KeyNSTREUWFTAOOKS-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point122°C to 125°C
Appearance (Form)Crystalline powder or needles
GC>=98.5 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only