1-Fluoro-2-nitrobenzene, 99%
1-Fluoro-2-nitrobenzene, 99%
1-Fluoro-2-nitrobenzene, 99%
Thermo Scientific Chemicals

1-Fluoro-2-nitrobenzene, 99%

CAS: 1493-27-2 | C6H4FNO2 | 141.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 119480100
Price (EUR)
21,40
Each
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
21,40
Each
Specifications
Assay Percent Range98.5% min. (GC)
Beilstein05, 241
CAS Max %100.0
CAS Min %98.5
CAS1493-27-2
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only