1-Heptanol, 98%
1-Heptanol, 98%
1-Heptanol, 98%
Thermo Scientific Chemicals

1-Heptanol, 98%

CAS: 111-70-6 | C7H16O | 116.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 120360025
Price (EUR)
104,00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
104,00
Each
Chemical Identifiers
CAS111-70-6
IUPAC Nameheptan-1-ol
Molecular FormulaC7H16O
InChI KeyBBMCTIGTTCKYKF-UHFFFAOYSA-N
SMILESCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Bromine number=<0.1 g Br/100 g
Infrared spectrumConforms
Acid number=<0.5 mg KOH/g
Residue after evaporation=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only