1-Indanone, 99+%
1-Indanone, 99+%
1-Indanone, 99+%
Thermo Scientific Chemicals

1-Indanone, 99+%

CAS: 83-33-0 | C9H8O | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Catalog number 122111000
Price (EUR)
237,00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
237,00
Each
Add to cart
Chemical Identifiers
CAS83-33-0
IUPAC Name2,3-dihydro-1H-inden-1-one
Molecular FormulaC9H8O
InChI KeyQNXSIUBBGPHDDE-UHFFFAOYSA-N
SMILESO=C1CCC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Infrared spectrumConforms
Melting point38°C to 42°C
Appearance (Color)Light yellow to brown
Appearance (Form)Crystalline mass
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only