Thermo Scientific Chemicals

Indene, 90%, tech., stabilized, Thermo Scientific Chemicals

Catalog number: 122142500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Indene, 90%, tech., stabilized, Thermo Scientific Chemicals

Catalog number: 122142500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 122142500
Price (EUR)
Price: 75,20
Online price: 67,68
Your price:
Quantity
-

Chemical Identifiers

CAS
95-13-6
IUPAC Name
1H-indene
Molecular Formula
C9H8
InChI Key
YBYIRNPNPLQARY-UHFFFAOYSA-N
SMILES
C1C=CC2=CC=CC=C12
Refractive index
1.5670 to 1.5750 (20°C, 589 nm)
Appearance (Color)
Clear yellow to brown
GC
>=88 %
Stabilizer
5 to 10 ppm tert-Butylcatechol
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 3
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Chronic aquatic toxicity
    Category 2
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H411 - Toxic to aquatic life with long lasting effects

    Physical Hazards

    H226 - Flammable liquid and vapor

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P332 + P313 - If skin irritation occurs: Get medical advice/attention

    P337 + P313 - If eye irritation persists: Get medical advice/attention