1-(Diphenylmethyl)piperazine, 97%
1-(Diphenylmethyl)piperazine, 97%
1-(Diphenylmethyl)piperazine, 97%
Thermo Scientific Chemicals

1-(Diphenylmethyl)piperazine, 97%

CAS: 841-77-0 | C17H20N2 | 252.361 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 122935000
Price (EUR)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS841-77-0
IUPAC Name1-(diphenylmethyl)piperazin-1-ium chloride
Molecular FormulaC17H21ClN2
InChI KeyDLCXUFKAJDKSDD-UHFFFAOYSA-N
SMILES[Cl-].C1C[NH+](CCN1)C(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point88°C to 93°C
GC>=96.0 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only