Hydrochloric acid, pure, fuming, 37% solution in water
Hydrochloric acid, pure, fuming, 37% solution in water
Hydrochloric acid, pure, fuming, 37% solution in water
Thermo Scientific Chemicals

Hydrochloric acid, pure, fuming, 37% solution in water

Hydrochloric acid, 35.0 to 39.0%, ClH, CAS Number-7647-01-0, 7732-18-5, acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt, 10L, 57.0 deg.C, CHEBI:17883 | CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
25 L
Packaging:
View all
Plastic drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 124620100
Price (EUR)
114,00
Each
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
114,00
Each
Chemical Identifiers
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear solution
Free chlorine=<10 ppm (Cl)
Sulfate (SO4)=<100 ppm
Color scale=<10 APHA
Lead (Pb)=<2 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only