Malononitrile, 99%
Malononitrile, 99%
Malononitrile, 99%
Thermo Scientific Chemicals

Malononitrile, 99%

CAS: 109-77-3 | C3H2N2 | 66.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 125275000
Price (EUR)
94,60
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
94,60
Each
Chemical Identifiers
CAS109-77-3
IUPAC Namepropanedinitrile
Molecular FormulaC3H2N2
InChI KeyCUONGYYJJVDODC-UHFFFAOYSA-N
SMILESN#CCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting crystalline mass
Melting point30°C to 34°C
Appearance (Color)White to yellow-brown
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only