Methanesulfonyl chloride, 99.5%
Methanesulfonyl chloride, 99.5%
Methanesulfonyl chloride, 99.5%
Thermo Scientific Chemicals

Methanesulfonyl chloride, 99.5%

CAS: 124-63-0 | CH3ClO2S | 114.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
500 mL
1 L
2.5 L
Packaging:
View all
Glass bottle
Catalog number 125645000
Price (EUR)
76,00
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
76,00
Each
Chemical Identifiers
CAS124-63-0
IUPAC Namemethanesulfonyl chloride
Molecular FormulaCH3ClO2S
InChI KeyQARBMVPHQWIHKH-UHFFFAOYSA-N
SMILESCS(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Color scale=<100 APHA
GC>=99.4 %
Refractive index1.4510 to 1.4530 (20°C, 589 nm)
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only