Methyl acetoacetate, 99+%
Methyl acetoacetate, 99+%
Methyl acetoacetate, 99+%
Thermo Scientific Chemicals

Methyl acetoacetate, 99+%

CAS: 105-45-3 | C5H8O3 | 116.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
View all
Plastic drum
Catalog number 126150100
Price (EUR)
329,00
Each
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
329,00
Each
Chemical Identifiers
CAS105-45-3
IUPAC Namemethyl 3-oxobutanoate
Molecular FormulaC5H8O3
InChI KeyWRQNANDWMGAFTP-UHFFFAOYSA-N
SMILESCOC(=O)CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Refractive index1.4175 to 1.4200 (20°C, 589 nm)
Free acid (CH3COOH)=<0.2 %
Color scale=<20 APHA
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only