4'-Methylacetophenone, 95%
4'-Methylacetophenone, 95%
4'-Methylacetophenone, 95%
Thermo Scientific Chemicals

4'-Methylacetophenone, 95%

CAS: 122-00-9 | C9H10O | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 126182500
Price (EUR)
37,10
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
37,10
Each
Chemical Identifiers
CAS122-00-9
IUPAC Name1-(4-methylphenyl)ethan-1-one
Molecular FormulaC9H10O
InChI KeyGNKZMNRKLCTJAY-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=94 %
Appearance (Color)Clear colorless to yellow
Refractive index1.5300 to 1.5350 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only