Methyl 4-(chloroformyl)butyrate, 98%
Methyl 4-(chloroformyl)butyrate, 98%
Methyl 4-(chloroformyl)butyrate, 98%
Methyl 4-(chloroformyl)butyrate, 98%
Thermo Scientific Chemicals

Methyl 4-(chloroformyl)butyrate, 98%

CAS: 1501-26-4 | C6H9ClO3 | 164.59 g/mol
Have Questions?
Catalog NumberQuantity
12654050050 g
Catalog number 126540500
Price (EUR)
182,00
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (EUR)
182,00
Each
Add to cart
Chemical Identifiers
CAS1501-26-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4440 to 1.4470 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only