3-Methyl-4-nitrobenzoic acid, 99%
3-Methyl-4-nitrobenzoic acid, 99%
3-Methyl-4-nitrobenzoic acid, 99%
Thermo Scientific Chemicals

3-Methyl-4-nitrobenzoic acid, 99%

CAS: 3113-71-1 | C8H7NO4 | 181.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 127285000
Price (EUR)
123,00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
123,00
Each
Chemical Identifiers
CAS3113-71-1
IUPAC Name3-methyl-4-nitrobenzoic acid
Molecular FormulaC8H7NO4
InChI KeyXDTTUTIFWDAMIX-UHFFFAOYSA-N
SMILESCC1=CC(=CC=C1[N+]([O-])=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Yellow
Melting point216°C to 219°C
Infrared spectrumConforms
Titration with NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only