6-Methyl-2-pyridinecarboxaldehyde, 97%
6-Methyl-2-pyridinecarboxaldehyde, 97%
6-Methyl-2-pyridinecarboxaldehyde, 97%
Thermo Scientific Chemicals

6-Methyl-2-pyridinecarboxaldehyde, 97%

CAS: 1122-72-1 | C7H7NO | 121.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 127570010
Price (EUR)
46,70
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
46,70
Each
Chemical Identifiers
CAS1122-72-1
IUPAC Name6-methylpyridine-2-carbaldehyde
Molecular FormulaC7H7NO
InChI KeyAHISYUZBWDSPQL-UHFFFAOYSA-N
SMILESCC1=CC=CC(C=O)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting solid
GC>=96.0 %
Appearance (Color)Light yellow to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only