N-Methylthiourea, 97%
N-Methylthiourea, 97%
N-Methylthiourea, 97%
Thermo Scientific Chemicals

N-Methylthiourea, 97%

CAS: 598-52-7 | C2H6N2S | 90.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 127900250
Price (EUR)
34,65
Online exclusive
38,40
Save 3,75 (10%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
34,65
Online exclusive
38,40
Save 3,75 (10%)
Each
Chemical Identifiers
CAS598-52-7
IUPAC Namemethylthiourea
Molecular FormulaC2H6N2S
InChI KeyKQJQICVXLJTWQD-UHFFFAOYSA-N
SMILESCNC(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point118°C to 123°C
Appearance (Form)Crystalline powder
HPLC>=96.0 %
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only