Norbornylene, 99%, stabilized
Norbornylene, 99%, stabilized
Norbornylene, 99%, stabilized
Thermo Scientific Chemicals

Norbornylene, 99%, stabilized

CAS: 498-66-8 | C7H10 | 94.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Packaging:
Glass bottle
Catalog number 129245000
Price (EUR)
79,00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
79,00
Each
Chemical Identifiers
CAS498-66-8
IUPAC Namebicyclo[2.2.1]hept-2-ene
Molecular FormulaC7H10
InChI KeyJFNLZVQOOSMTJK-UHFFFAOYNA-N
SMILESC1CC2CC1C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Antioxidant0.1 % Phenolic antioxidant
Antioxidant0.1 % Phenolic antioxidant
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only