Norharman, 98%
Norharman, 98%
Norharman, 98%
Thermo Scientific Chemicals

Norharman, 98%

CAS: 244-63-3 | C11H8N2 | 168.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 129260025
Price (EUR)
523,65
Online exclusive
582,00
Save 58,35 (10%)
Each
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
523,65
Online exclusive
582,00
Save 58,35 (10%)
Each
Chemical Identifiers
CAS244-63-3
IUPAC Name9H-pyrido[3,4-b]indole
Molecular FormulaC11H8N2
InChI KeyAIFRHYZBTHREPW-UHFFFAOYSA-N
SMILESN1C2=C(C=CC=C2)C2=C1C=NC=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light beige
Appearance (Form)Powder
Melting point198°C to 202°C
Titration Tertiary base>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only