Octadecylamine, 90%, tech.
Octadecylamine, 90%, tech.
Octadecylamine, 90%, tech.
Thermo Scientific Chemicals

Octadecylamine, 90%, tech.

CAS: 124-30-1 | C18H39N | 269.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number 129320050
Price (EUR)
402,00
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
402,00
Each
Chemical Identifiers
CAS124-30-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Pastilles
Infrared spectrumConforms
Appearance (Color)White to cream
GC>=88 %
Amine number204 to 211 mg KOH/g (Total)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only