Phenanthrene, 97%
Phenanthrene, 97%
Phenanthrene, 97%
Thermo Scientific Chemicals

Phenanthrene, 97%

CAS: 85-01-8 | C14H10 | 178.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
500 g
Packaging:
View all
Glass bottle
Catalog number 130090050
Price (EUR)
21,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
21,00
Each
Chemical Identifiers
CAS85-01-8
IUPAC Namephenanthrene
Molecular FormulaC14H10
InChI KeyYNPNZTXNASCQKK-UHFFFAOYSA-N
SMILESC1=CC=C2C(C=CC3=CC=CC=C23)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White to grey to light brown
Infrared spectrumConforms
GC>=96.0 %
Melting point97°C to 101°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only