2-Phenylphenol, 99+%
2-Phenylphenol, 99+%
2-Phenylphenol, 99+%
Thermo Scientific Chemicals

2-Phenylphenol, 99+%

CAS: 90-43-7 | C12H10O | 170.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 130760050
Price (EUR)
27,10
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
27,10
Each
Chemical Identifiers
CAS90-43-7
IUPAC Name[1,1'-biphenyl]-2-ol
Molecular FormulaC12H10O
InChI KeyLLEMOWNGBBNAJR-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
GC>=99.0 %
Appearance (Form)Crystalline flakes
Melting point56°C to 59°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only