Piperazine, 99%, extra pure
Piperazine, 99%, extra pure
Piperazine, 99%, extra pure
Thermo Scientific Chemicals

Piperazine, 99%, extra pure

CAS: 110-85-0 | C4H10N2 | 86.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
10 kg
Catalog number 131295000
Price (EUR)
60,65
Online Exclusive
67,20
Save 6,55 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
60,65
Online Exclusive
67,20
Save 6,55 (10%)
Each
Chemical Identifiers
CAS110-85-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline flakes
Color of solution=<30 APHA (50% in methanol)
Appearance (Color)White to light yellow
Infrared spectrumConforms
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only