Piperonylic acid, 99%
Piperonylic acid, 99%
Piperonylic acid, 99%
Thermo Scientific Chemicals

Piperonylic acid, 99%

CAS: 94-53-1 | C8H6O4 | 166.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Plastic bottle
Catalog number 131371000
Price (EUR)
212,00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
212,00
Each
Chemical Identifiers
CAS94-53-1
IUPAC Name2H-1,3-benzodioxole-5-carboxylic acid
Molecular FormulaC8H6O4
InChI KeyVDVJGIYXDVPQLP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Appearance (Color)Off-white to beige
Titration with NaOH>=98.5 %
Melting point229°C to 235°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only