Propargyl alcohol, 99%
Propargyl alcohol, 99%
Propargyl alcohol, 99%
Thermo Scientific Chemicals

Propargyl alcohol, 99%

CAS: 107-19-7 | C3H4O | 56.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 131455000
Price (EUR)
57,80
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
57,80
Each
Chemical Identifiers
CAS107-19-7
IUPAC Nameprop-2-yn-1-ol
Molecular FormulaC3H4O
InChI KeyTVDSBUOJIPERQY-UHFFFAOYSA-N
SMILESOCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.4300 to 1.4330 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only