Propionyl chloride, 98%
Propionyl chloride, 98%
Propionyl chloride, 98%
Thermo Scientific Chemicals

Propionyl chloride, 98%

CAS: 79-03-8 | C3H5ClO | 92.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
500 g
2.5 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 131532500
Price (EUR)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS79-03-8
IUPAC Namepropanoyl chloride
Molecular FormulaC3H5ClO
InChI KeyRZWZRACFZGVKFM-UHFFFAOYSA-N
SMILESCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration RCOCl>=97.5 %
GC>=97.5 %
Appearance (Form)Clear liquid
Color scale=<20 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only