Pyrazole, 98%, pure
Pyrazole, 98%, pure
Pyrazole, 98%, pure
Thermo Scientific Chemicals

Pyrazole, 98%, pure

CAS: 288-13-1 | C3H4N2 | 68.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 131740250
Price (EUR)
47,65
キャンペーン価格
52,80
Save 5,15 (10%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
47,65
キャンペーン価格
52,80
Save 5,15 (10%)
Each
Chemical Identifiers
CAS288-13-1
IUPAC Name1H-pyrazole
Molecular FormulaC3H4N2
InChI KeyWTKZEGDFNFYCGP-UHFFFAOYSA-N
SMILESN1C=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point66°C to 70°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only