Quinoline, 96%
Quinoline, 96%
Quinoline, 96%
Thermo Scientific Chemicals

Quinoline, 96%

CAS: 91-22-5 | C9H7N | 129.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
10 L
Packaging:
View all
Plastic drum
Catalog number 132190100
Price (EUR)
574,65
Online exclusive
638,00
Save 63,35 (10%)
Each
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
574,65
Online exclusive
638,00
Save 63,35 (10%)
Each
Chemical Identifiers
CAS91-22-5
IUPAC Namequinoline
Molecular FormulaC9H7N
InChI KeySMWDFEZZVXVKRB-UHFFFAOYSA-N
SMILESC1=CC=C2N=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow to dark brown
Infrared spectrumConforms
Refractive index1.6230 to 1.6270 (20°C, 589 nm)
GC>=95 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only