Sebacic Acid, 98%
Sebacic Acid, 98%
Sebacic Acid, 98%
Thermo Scientific Chemicals

Sebacic Acid, 98%

CAS: 111-20-6 | C10H18O4 | 202.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 132630025
Price (EUR)
254,00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
254,00
Each
Chemical Identifiers
CAS111-20-6
IUPAC Namedecanedioic acid
Molecular FormulaC10H18O4
InChI KeyCXMXRPHRNRROMY-UHFFFAOYSA-N
SMILESOC(=O)CCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or granules
GC>=97.5 %
Appearance (Color)White to off-white
Infrared spectrumConforms
Ash=<0.06 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only