beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol
Thermo Scientific Chemicals

beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol

CAS: 83-46-5 | C29H50O | 414.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Catalog number 132721000
Price (EUR)
95,10
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
95,10
Each
Chemical Identifiers
CAS83-46-5
IUPAC Name(1R,3aS,3bS,7S,9aR,9bS,11aR)-1-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Molecular FormulaC29H50O
InChI KeyKZJWDPNRJALLNS-VJSFXXLFSA-N
SMILESCC[C@H](CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<3.5 %
GC>=95 % Total sterols
GC10 to 14 % (Beta-sitostanol) Typical
Appearance (Form)Powder
Specific optical rotation-24° to -32° (20°C, 589 nm) (c=2, CHCl3)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only