Spermine, 97%
Spermine, 97%
Spermine, 97%
Thermo Scientific Chemicals

Spermine, 97%

CAS: 71-44-3 | C10H26N4 | 202.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 132750010
Price (EUR)
51,65
線上優惠
57,70
Save 6,05 (10%)
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
51,65
線上優惠
57,70
Save 6,05 (10%)
Each
Chemical Identifiers
CAS71-44-3
IUPAC Name(3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine
Molecular FormulaC10H26N4
InChI KeyPFNFFQXMRSDOHW-UHFFFAOYSA-N
SMILESNCCCNCCCCNCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<3 % (K.F.)
Appearance (Color)White to light yellow
Melting point26°C to 30°C
Appearance (Form)Crystalline powder or low melting mass
Titration with HClO4>=96.0 % (On dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only