Styrene oxide, 97+%
Styrene oxide, 97+%
Styrene oxide, 97+%
Thermo Scientific Chemicals

Styrene oxide, 97+%

CAS: 96-09-3 | C8H8O | 120.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
250 mL
1 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 132800100
Price (EUR)
24,80
Each
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
24,80
Each
Chemical Identifiers
CAS96-09-3
IUPAC Name2-phenyloxirane
Molecular FormulaC8H8O
InChI KeyAWMVMTVKBNGEAK-UHFFFAOYNA-N
SMILESC1OC1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5330 to 1.5360 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
GC>=97.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only