3,3,5,5-Tetramethylcyclohexanone, 98%
3,3,5,5-Tetramethylcyclohexanone, 98%
3,3,5,5-Tetramethylcyclohexanone, 98%
Thermo Scientific Chemicals

3,3,5,5-Tetramethylcyclohexanone, 98%

CAS: 14376-79-5 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Catalog number 138390500
Price (EUR)
277,00
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
277,00
Each
Chemical Identifiers
CAS14376-79-5
IUPAC Name3,3,5,5-tetramethylcyclohexan-1-one
Molecular FormulaC10H18O
InChI KeyOQJMHUOCLRCSED-UHFFFAOYSA-N
SMILESCC1(C)CC(=O)CC(C)(C)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4510 to 1.4530 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only