Tetramethylthiuram disulfide, 97%
Tetramethylthiuram disulfide, 97%
Tetramethylthiuram disulfide, 97%
Tetramethylthiuram disulfide, 97%
Thermo Scientific Chemicals

Tetramethylthiuram disulfide, 97%

CAS: 137-26-8 | C6H12N2S4 | 240.416 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
1 kg
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 138485000
Price (EUR)
79,20
Each
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
79,20
Each
Add to cart
Chemical Identifiers
CAS137-26-8
IUPAC NameN,N-dimethyl[(dimethylcarbamothioyl)disulfanyl]carbothioamide
Molecular FormulaC6H12N2S4
InChI KeyKUAZQDVKQLNFPE-UHFFFAOYSA-N
SMILESCN(C)C(=S)SSC(=S)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Infrared spectrumConforms
HPLC>=96.0 %
Ash=<0.5 %
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only