o-Toluidine, 99%
o-Toluidine, 99%
o-Toluidine, 99%
Thermo Scientific Chemicals

o-Toluidine, 99%

CAS: 95-53-4 | C7H9N | 107.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
10 L
Packaging:
View all
Plastic drum
Catalog number 139080100
Price (EUR)
472,00
Each
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
472,00
Each
Chemical Identifiers
CAS95-53-4
IUPAC Name2-methylaniline
Molecular FormulaC7H9N
InChI KeyRNVCVTLRINQCPJ-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5710 to 1.5730 (20°C, 589 nm)
Appearance (Color)Clear yellow to orange to light brown
AppearanceMay darken on storage
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only