Trifluoroacetic acid, 99+%, extra pure
Trifluoroacetic acid, 99+%, extra pure
Trifluoroacetic acid, 99+%, extra pure
Thermo Scientific Chemicals

Trifluoroacetic acid, 99+%, extra pure

CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Glass Bottle
Catalog number 139720010
Price (EUR)
390,00
Each
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
390,00
Each
Chemical Identifiers
CAS76-05-1
IUPAC Nametrifluoroacetic acid
Molecular FormulaC2HF3O2
InChI KeyDTQVDTLACAAQTR-UHFFFAOYSA-N
SMILESOC(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.3 % (Coulometric)
Color scale=<20 APHA
Appearance (Form)Clear liquid
Titration with NaOH99.0 to 101.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only