Trimethylacetonitrile, 98%
Trimethylacetonitrile, 98%
Trimethylacetonitrile, 98%
Thermo Scientific Chemicals

Trimethylacetonitrile, 98%

CAS: 630-18-2 | C5H9N | 83.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 140031000
Price (EUR)
166,00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
166,00
Each
Chemical Identifiers
CAS630-18-2
IUPAC Name2,2-dimethylpropanenitrile
Molecular FormulaC5H9N
InChI KeyJAMNHZBIQDNHMM-UHFFFAOYSA-N
SMILESCC(C)(C)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.3770 to 1.3790 (20°C, 589 nm)
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only