o-Xylene, 99%, pure
o-Xylene, 99%, pure
o-Xylene, 99%, pure
Thermo Scientific Chemicals

o-Xylene, 99%, pure

CAS: 95-47-6 | C8H10 | 106.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 140990010
Price (EUR)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS95-47-6
IUPAC Name1,2-xylene
Molecular FormulaC8H10
InChI KeyCTQNGGLPUBDAKN-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.5035 to 1.5065 (20°C, 589 nm)
GC>=98.5 %
Residue after evaporation=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only