m-Xylylenediamine, 99%
m-Xylylenediamine, 99%
m-Xylylenediamine, 99%
Thermo Scientific Chemicals

m-Xylylenediamine, 99%

CAS: 1477-55-0 | C8H12N2 | 136.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 141025000
Price (EUR)
130,00
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
130,00
Each
Chemical Identifiers
CAS1477-55-0
IUPAC Name1-[3-(aminomethyl)phenyl]methanamine
Molecular FormulaC8H12N2
InChI KeyFDLQZKYLHJJBHD-UHFFFAOYSA-N
SMILESNCC1=CC(CN)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with HClO4>=98.5 %
Refractive index1.5700 to 1.5720 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only