Adamantane, 99+%
Adamantane, 99+%
Adamantane, 99+%
Thermo Scientific Chemicals

Adamantane, 99+%

CAS: 281-23-2 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number 146110010
Price (EUR)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS281-23-2
IUPAC Nameadamantane
Molecular FormulaC10H16
InChI KeyORILYTVJVMAKLC-UHFFFAOYSA-N
SMILESC1C2CC3CC1CC(C2)C3
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Appearance (Color)White to beige
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only