Para Red, pure
Para Red, pure
Para Red, pure
Thermo Scientific Chemicals

Para Red, pure

C16H11N3O3
Have Questions?
Catalog NumberQuantity
14630025025 g
Catalog number 146300250
Price (EUR)
104,65
線上優惠
116,00
Save 11,35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
104,65
線上優惠
116,00
Save 11,35 (10%)
Each
Chemical Identifiers
CAS6410-10-2
IUPAC Name(1Z)-1-[2-(4-nitrophenyl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
Molecular FormulaC16H11N3O3
InChI KeyACHQDMGGRZPFAW-VLGSPTGOSA-N
SMILES[O-][N+](=O)C1=CC=C(NN=C2/C(=O)C=CC3=CC=CC=C23)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only