4-Heptanone, 98%
4-Heptanone, 98%
4-Heptanone, 98%
Thermo Scientific Chemicals

4-Heptanone, 98%

CAS: 123-19-3 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 146550050
Price (EUR)
24,10
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
24,10
Each
Chemical Identifiers
CAS123-19-3
IUPAC Nameheptan-4-one
Molecular FormulaC7H14O
InChI KeyHCFAJYNVAYBARA-UHFFFAOYSA-N
SMILESCCCC(=O)CCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.4060 to 1.4080 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only