Ethyl phenylacetate, 99%
Product Image
Thermo Scientific Chemicals

Ethyl phenylacetate, 99%

CAS: 101-97-3 | C10H12O2 | 164.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Catalog number 148430010
Price (EUR)
221,00
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
221,00
Each
Add to cart
Chemical Identifiers
CAS101-97-3
IUPAC Nameethyl 2-phenylacetate
Molecular FormulaC10H12O2
InChI KeyDULCUDSUACXJJC-UHFFFAOYSA-N
SMILESCCOC(=O)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Acid value=<1 mg KOH/g
GC>=98.5 %
Refractive index1.4970 to 1.4990 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only